Anti-CES1 Polyclonal Antibody

Lieferant: Boster Bio
A01741-1
BSBTA01741-1EA 696 CHF
BSBTA01741-1
Anti-CES1 Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for CES1 detection. Tested with WB, IHC-P, Direct ELISA in Human;Mouse;Rat.

Type: Primary
Antigen: CES1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR