Anti-VCL Rabbit Polyclonal Antibody

Lieferant: Boster Bio
A01207
BSBTA01207EA 696 CHF
BSBTA01207
Anti-VCL Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Vinculin(VCL) detection. Tested with WB in Human.

Type: Primary
Antigen: VCL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR