Anti-MUT Rabbit Polyclonal Antibody

Lieferant: Boster Bio
A01065
BSBTA01065EA 696 CHF
BSBTA01065
Anti-MUT Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Methylmalonyl-CoA mutase, mitochondrial(MUT) detection. Tested with WB, IHC-P in Human, Rat.

Type: Primary
Antigen: MUT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR