Anti-CAST Rabbit Polyclonal Antibody

Lieferant: Boster Bio
A00337
BSBTA00337EA 696 CHF
BSBTA00337
Anti-CAST Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Calpastatin(CAST) detection. Tested with WB, IHC-P in Human.

Type: Primary
Antigen: CAST
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR