Labels for BBP33 printers, type B-425, WorkHorse™
Lieferant: Brady
142891 143053 142962 142892 361734 361733
BRDY142891EA 157 CHF
BRDY142891 BRDY143053 BRDY142962 BRDY142892 BRDY361734 BRDY361733
Labels for BBP33 printers, type B-425, WorkHorse™
Etiketten
These polypropylene labels have a matt finish and are used for product and general purpose identification, general labeling for indoor use that requires outstanding chemical and solvent resistance.