Anti-CGI62/C8orf70 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-9666R-A680
BOSSBS-9666R-A680EA 1190 CHF
BOSSBS-9666R-A680
Anti-CGI62/C8orf70 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Made up of nearly 146 million bases, chromosome 8 encodes about 800 genes. Translocation of portions of chromosome 8 with amplifications of the c-Myc gene are found in some leukaemias and lymphomas, and typically associated with a poor prognosis. Portions of chromosome 8 have been linked to schizophrenia and bipolar disorder. Trisomy 8, also known as Warkany syndrome 2, most often results in early miscarriage but is occasionally seen in a mosaic form in surviving patients who suffer to a varying degree from a number of symptoms including retarded mental and motor development, and certainMore Product Information
Order Now

TECHNISCHE DATEN

Environmentally Preferable
Antigen-Name CGI62/C8orf70
Antigen-Synonyme C8orf70|fam164a|CGI-62|hypothetical protein LOC51101|CGI 62|F164A_HUMAN|Hypothetical protein C8orf70|Chromosome 8 open reading frame 70|family with sequence similarity 164, member A|Protein FAM164A
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Alexa Fluor® 680
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 4879
Isotyp IgG
Western blot Yes
ImmunoChemie Yes
ImmunFluoreszenz Yes
Modification Unmodified
Epitop 101-200/325
Immunogen KLH conjugated synthetic peptide derived from human C8orf70
Form Liquid
Reinigung Purified by Protein A
Konzentration 1 µg/µl
Lagerungspuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagertemperatur Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Transporttemperatur 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR