Anti-TEF3 Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-9603R-CY3
BOSSBS-9603R-CY3EA 1200 CHF
BOSSBS-9603R-CY3
Anti-TEF3 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
TEF-3, is a 427 amino acid member of the transcriptional enhancer factor (TEF) family of proteins that are characterized by the presence of a TEA DNA-binding domain. Localized to the nucleus and expressed primarily in skeletal muscle, TEF-3 functions as a transcriptional regulator by binding specifically and non-cooperatively to the M-CAT motif found in the promotors of muscle-specific genes, thereby directing their subsequent expression. TEF-3 contains one TEA DNA-binding domain and is expressed as multiple isoforms due to alternative splicing events.
Recommended Dilutions: IF(IHC-P): 1:5More Product Information