Anti-GDPD3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-9206R-A680
BOSSBS-9206R-A680EA 1190 CHF
BOSSBS-9206R-A680
Anti-GDPD3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Hydrolyzes lysoglycerophospholipids to produce lysophosphatidic acid (LPA) and the corresponding amines. Shows a preference for 1-O-alkyl-sn-glycero-3-phosphocholine (lyso-PAF), lysophosphatidylcholine (lyso-PC) and N-acylethanolamine lysophospholipids. Does not display glycerophosphodiester phosphodiesterase activity, since it cannot hydrolyze either glycerophosphoinositol or glycerophosphocholine.gene mapping to human chromosome Xq13.1.

Type: Primary
Antigen: GDPD3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/318
Host: Rabbit
Isotype: IgG
More Product Information
Order Now

TECHNISCHE DATEN

Antigen-Name GDPD3
Antigen-Synonyme GDPD3|GDE7|Glycerophosphodiester phosphodiesterase domain-containing protein 3
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Alexa Fluor® 680
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 79153
Isotyp IgG
Western blot Yes
ImmunoChemie Yes
ImmunFluoreszenz Yes
Modification Unmodified
Epitop 101-200/318
Immunogen KLH conjugated synthetic peptide derived from human GDPD3
Form Liquid
Molecular weight 37 kDa
Reinigung Purified by Protein A
Konzentration 1 µg/µl
Lagerungspuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagertemperatur Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Transporttemperatur 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR