Anti-VGLL1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-9182R-HRP
BOSSBS-9182R-HRPEA 1200 CHF
BOSSBS-9182R-HRP
Anti-VGLL1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
VGLL1 is a specific coactivator for the mammalian TEFs. The mammalian TEF and the Drosophila scalloped genes belong to a conserved family of transcriptional factors that possesses a TEA/ATTS DNA binding domain.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: VGLL1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR