Anti-HPRT1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-9026R-A555
BOSSBS-9026R-A555EA 1190 CHF
BOSSBS-9026R-A555
Anti-HPRT1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
The protein encoded by this gene is a transferase, which catalyzes conversion of hypoxanthine to inosine monophosphate and guanine to guanosine monophosphate via transfer of the 5-phosphoribosyl group from 5-phosphoribosyl 1-pyrophosphate. This enzyme plays a central role in the generation of purine nucleotides through the purine salvage pathway. Mutations in this gene result in Lesch-Nyhan syndrome or gout.[provided by RefSeq, Jun 2009].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HPRT1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR