Anti-Viperin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-8718R-A750
BOSSBS-8718R-A750EA 1190 CHF
BOSSBS-8718R-A750
Anti-Viperin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
RSAD2 is a 361 amino acid protein that is involved in antiviral defense against pathogens such as Hep C, Cytomegalovirus and HIV-1. Localised to the cytosolic side of the endoplasmic reticulum and relocated to the Golgi apparatus upon viral infection, RSAD2 is thought to prevent viral budding by disrupting lipid rafts at the plasma membrane and supporting the Interferon-induced antiviral state of the cell. Additionally, RSAD2 can bind to and inactivate FDPS (an enzyme that is crucial for the synthesis of cholesterol and geranylated and farnesylated proteins), thereby playing a role in lipidMore Product Information