Anti-RNASE3 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-8615R-CY5
BOSSBS-8615R-CY5EA 1200 CHF
BOSSBS-8615R-CY5
Anti-RNASE3 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Cytotoxin and helminthotoxin with low-efficiency ribonuclease activity. Possesses a wide variety of biological activities. Exhibits antibacterial activity, including cytoplasmic membrane depolarization of preferentially Gram-negative, but also Gram-positive strains. Promotes E.coli outer membrane detachment, alteration of the overall cell shape and partial loss of cell content.

Recommended Dilutions: IF(IHC-P): 1:50-200; IF(IHC-F)(1:50-200

Type: Primary
Antigen: RNASE3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

TECHNISCHE DATEN

Antigen-Symbol RNASE3
Environmentally Preferable
Antigen-Name Ribonuclease A Family Member 3
Antigen-Synonyme RNS3|ECP
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Cy5®
Reaktivität Human
Wirt Rabbit
Gen-ID 6037
Isotyp IgG
ImmunoChemie Yes
Immunogen 40-90/160
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagertemperatur Store at –20 °C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR