Anti-UBE2L6 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-8369R
BOSSBS-8369REA 965 CHF
BOSSBS-8369R
Anti-UBE2L6 Rabbit Polyclonal Antibody
Antikörper
Ubiquitin is an abundant, highly conserved protein found in all eukaryotic cells, either free or covalently attached to cellular proteins. The primary function of ubiquitin in mammalian systems is to clear abnormal, foreign, and improperly folded proteins by targeting them for proteosome degradation. Ubiquitin conjugating enzyme 8 (UBC8) is an E2 enzyme involved in the ubiquitin pathway for protein degradation. Like other E2 enzymes, UBC8 forms a thioester bond with ubiquitin in an E1-dependent manner. UBC8 binds to the human homolog of Drosophila ariadne (HHARI) and UBC7-associated protein (More Product Information
Order Now

TECHNISCHE DATEN

Antigen-Symbol UBE2L6
Environmentally Preferable
Antigen-Name Ubiquitin-conjugating enzyme E2L 6
Antigen-Synonyme UbcH8|RIG-B|Retinoic acid-induced gene B protein|Ubiquitin/ISG15-conjugating enzyme E2 L6|Ubiquitin carrier protein L6|Ubiquitin-protein ligase L6
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 9246
Isotyp IgG
Western blot Yes
ImmunoChemie Yes
Form liquid
Reinigung Purified by Protein A.
Konzentration 1 ug/ul
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Transporttemperatur 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR