Anti-HCV NS5A transactivated protein 8 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-8158R-A680
BOSSBS-8158R-A680EA 1190 CHF
BOSSBS-8158R-A680
Anti-HCV NS5A transactivated protein 8 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
BTBD1 contains a proline-rich region and a BTB/POZ domain which are typically involved in protein-protein interactions. The C-terminus of BTBD1 binds topoisomerase I and BTBD1 also interacts with TRIM5 isoform Delta. BTBD1 localises to cytoplasmic bodies and is essential for myogenic differentiation.

Type: Primary
Antigen: HCV NS5A transactivated protein 8
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 401-482/482
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

TECHNISCHE DATEN

Antigen-Name HCV NS5A transactivated protein 8
Environmentally Preferable
Antigen-Synonyme C15orf1|Hepatitis C virus NS5A transactivated protein 8|BTB domain containing 1|BTB POZ domain containing 1|BTBD1_HUMAN|HCV NS5A transactivated protein 8|BTB/POZ domain containing protein 1|NS5ATP8|BTBD 1
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Alexa Fluor® 680
Reaktivität
Wirt Rabbit
Gen-ID 53339
Isotyp IgG
Western blot Yes
ImmunoChemie Yes
ImmunFluoreszenz Yes
Modification Unmodified
Epitop 401-482/482
Immunogen KLH conjugated synthetic peptide derived from human BTBD1/HCV NS5A transactivated protein 8
Form Liquid
Molecular weight 53 kDa
Reinigung Purified by Protein A
Konzentration 1 µg/µl
Lagerungspuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagertemperatur Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Transporttemperatur 4 °C
Validierte Anwendungen ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR