Anti-PLEKHM3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BS-8064R-A488
BOSSBS-8064R-A488EA 1190 CHF
BOSSBS-8064R-A488
Anti-PLEKHM3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
PLEKHM3 (pleckstrin homology domain containing, family M, member 3), also known as DAPR or PLEKHM1L (pleckstrin homology domain containing, family M, member 1-like), is a 761 amino acid phosphoprotein that contains two pleckstrin homology (PH) domains and one phorbol-ester/DAG-type zinc finger. Conserved in chimpanzee, dog, cow, mouse, rat, chicken and zebrafish, PLEKHM3 exists as three alternatively spliced isoforms that participate in metal ion binding. The gene that encodes PLEKHM3 maps to human chromosome 2q33.3. As the second largest human chromosome, chromosome 2 makes up approximatelyMore Product Information