Anti-DNPH1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-7827R-A350
BOSSBS-7827R-A350EA 1190 CHF
BOSSBS-7827R-A350
Anti-DNPH1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Catalyzes the cleavage of the N-glycosidic bond of deoxyribonucleoside 5'-monophosphates to yield deoxyribose 5-phosphate and a purine or pyrimidine base. Deoxyribonucleoside 5'-monophosphates containing purine bases are preferred to those containing pyrimidine bases.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DNPH1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR