Anti-ASB3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-7736R-HRP
BOSSBS-7736R-HRPEA 1200 CHF
BOSSBS-7736R-HRP
Anti-ASB3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Members of the suppressor of cytokine signaling (SOCS) family of proteins contain C-terminal regions of homology called the SOCS box, which serves to couple SOCS proteins and their binding partners with the elongin B and C complex. Serveral other families of proteins also contain SOCS boxes but differ from the SOCS proteins in the type of domain they contained upstream of the SOCS box. Four members of the ankyrin repeat and SOCS box-containing (ASB) protein family are identified and termed as ASB-1, ASB-2, ASB-3 and ASB-4. ASB-1 is expressed in multiple organs, including the hematopoieticMore Product Information
Order Now

TECHNISCHE DATEN

Antigen-Symbol ASB3
Environmentally Preferable
Antigen-Name Ankyrin Repeat And SOCS Box Containing 3
Antigen-Synonyme ASB3_HUMAN|Weakly similar to ankyrin brain variant 2.|ASB-3|Ankyrin repeat and SOCS box containing 3|ASB3
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation HRP (Horseradish Peroxidase)
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 51130
Isotyp IgG
Western blot Yes
ImmunoChemie Yes
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
Lagertemperatur Store at –20 °C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR