Anti-STAMBP Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-7735R-CY5
BOSSBS-7735R-CY5EA 1200 CHF
BOSSBS-7735R-CY5
Anti-STAMBP Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Zinc metalloprotease that specifically cleaves 'Lys-63'-linked polyubiquitin chains. Does not cleave 'Lys-48'-linked polyubiquitin chains (By similarity). Functions at the endosome and is able to oppose the ubiquitin-dependent sorting of receptors to lysosomes. Plays a role in signal transduction for cell growth and MYC induction mediated by IL-2 and GM-CSF. Potentiates BMP (bone morphogenetic protein) signaling by antagonizing the inhibitory action of SMAD6 and SMAD7.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: STAMBP
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR