Anti-MAGEA10 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-6816R-A680
BOSSBS-6816R-A680EA 1190 CHF
BOSSBS-6816R-A680
Anti-MAGEA10 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. Alternative splicing results in multiple transcript variants. Read-through transcription alsoMore Product Information
Order Now

TECHNISCHE DATEN

Antigen-Name MAGEA10
Environmentally Preferable
Antigen-Synonyme Cancer/testis antigen 1.10|Melanoma antigen family A 10|CT1.10|MAGE 10 antigen|MAGAA_HUMAN|MAGE10|Melanoma associated antigen 10
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Alexa Fluor® 680
Reaktivität
Wirt Rabbit
Gen-ID 4109
Isotyp IgG
ImmunoChemie Yes
ImmunFluoreszenz Yes
Modification Unmodified
Epitop 241-340/369
Immunogen KLH conjugated synthetic peptide derived from human MAGEA10
Form Liquid
Molecular weight 41 kDa
Reinigung Purified by Protein A
Konzentration 1 µg/µl
Lagerungspuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagertemperatur Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Transporttemperatur 4 °C
Validierte Anwendungen ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR