Anti-UGDH Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-6715R-A680
BOSSBS-6715R-A680EA 1190 CHF
BOSSBS-6715R-A680
Anti-UGDH Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
The protein encoded by this gene converts UDP-glucose to UDP-glucuronate and thereby participates in the biosynthesis of glycosaminoglycans such as hyaluronan, chondroitin sulfate, and heparan sulfate. These glycosylated compounds are common components of the extracellular matrix and likely play roles in signal transduction, cell migration, and cancer growth and metastasis. The expression of this gene is up-regulated by transforming growth factor beta and down-regulated by hypoxia. Alternative splicing results in multiple transcript variants.[provided by RefSeq, May 2010].
Type: Primary