Anti-IHH Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-6624R-CY5.5
BOSSBS-6624R-CY5.5EA 1200 CHF
BOSSBS-6624R-CY5.5
Anti-IHH Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Ihh is an intercellular signalling peptide essential for a variety of patterning events during development. It binds to the patched (PTC) receptor, which functions in association with smoothened (SMO), to activate the transcription of target genes. Ihh induces the expression of parathyroid hormone-related protein (PTHRP) and is implicated in endochondral ossification, possibly regulating the balance between growth and ossification of the developing bones.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IHH
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host:More Product Information
Order Now

TECHNISCHE DATEN

Antigen-Symbol IHH
Environmentally Preferable
Antigen-Name Indian Hedgehog Homolog
Antigen-Synonyme BDA1|HHG-2|HHG2
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Cy5.5®
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 3549
Isotyp IgG
ImmunoChemie Yes
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagertemperatur Store at 4°C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR