Anti-CHRNA3 Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-6455R-CY3
BOSSBS-6455R-CY3EA 1200 CHF
BOSSBS-6455R-CY3
Anti-CHRNA3 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Members of the ligand-gated ion channel receptor family are characterized by their fast transmitting response to neurotransmitters. Two important members of this family are the nicotinic acetylcholine and glutamate receptors, both of which are composed of five homologous subunits forming a transmembrane aqueous pore. These transmembrane receptors change conformation in response to their cognate neurotransmitter. Nicotinic acetylcholine receptors (AChRs) are found at the postsynaptic membrane of the neuromuscular junction and bind acetylcholine molecules, allowing ions to move through theMore Product Information