Anti-Quiescin Q6 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-6394R-CY7
BOSSBS-6394R-CY7EA 1200 CHF
BOSSBS-6394R-CY7
Anti-Quiescin Q6 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
This gene encodes a protein that contains domains of thioredoxin and ERV1, members of two long-standing gene families. The gene expression is induced as fibroblasts begin to exit the proliferative cycle and enter quiescence, suggesting that this gene plays an important role in growth regulation. Two transcript variants encoding two different isoforms have been found for this gene.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Quiescin Q6
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR