Anti-ADSL Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-6352R-CY3
BOSSBS-6352R-CY3EA 1200 CHF
BOSSBS-6352R-CY3
Anti-ADSL Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Adenylsuccinate lyase is involved in both de novo synthesis of purines and formation of adenosine monophosphate from inosine monophosphate. It catalyzes two reactions in AMP biosynthesis: the removal of a fumarate from succinylaminoimidazole carboxamide (SAICA) ribotide to give aminoimidazole carboxamide ribotide (AICA) and removal of fumarate from adenylosuccinate to give AMP. Adenylosuccinase deficiency results in succinylpurinemic autism, psychomotor retardation, and , in some cases, growth retardation associated with muscle wasting and epilepsy. Two transcript variants encoding differentMore Product Information