Anti-ESRRG Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-6213R
BOSSBS-6213REA 965 CHF
BOSSBS-6213R
Anti-ESRRG Rabbit Polyclonal Antibody
Antikörper
This gene encodes a member of the estrogen receptor-related receptor (ESRR) family, which belongs to the nuclear hormone receptor superfamily. All members of the ESRR family share an almost identical DNA binding domain, which is composed of two C4-type zinc finger motifs. The ESRR members are orphan nuclear receptors; they bind to the estrogen response element and steroidogenic factor 1 response element, and activate genes controlled by both response elements in the absence of any ligands. The ESRR family is closely related to the estrogen receptor (ER) family. They share target genes, co-More Product Information