Anti-SNF2L Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-6015R-A647
BOSSBS-6015R-A647EA 1190 CHF
BOSSBS-6015R-A647
Anti-SNF2L Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
SNF2L, also known as SMARCA1 (SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 1), SWI or ISWI, is a 1,054 amino acid protein that localizes to the nucleus and contains one helicase C-terminal domain, one helicase ATP-binding domain and two SANT domains. Expressed as multiple alternatively spliced isoforms, SNF2L exists as a component of the nucleosome-remodeling factor (NURF) complex where it helps to facilitate the ATP-dependent perturbation of chromatin structure and may also be involved in brain development and neurite outgrowth. The geneMore Product Information
Order Now

TECHNISCHE DATEN

Antigen-Symbol SNF2L
Antigen-Name SWI/SNF related matrix associated actin dependent Regulator of chromatin subfamily a member 1
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Alexa Fluor® 647
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 6594
Isotyp IgG
ImmunoChemie Yes
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagertemperatur Store at 4°C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR