Anti-PRODH Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-5813R-A680
BOSSBS-5813R-A680EA 1190 CHF
BOSSBS-5813R-A680
Anti-PRODH Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Proline oxidase catalyses the conversion of proline to pyrroline-5-carboxylate, or P5C during the degradation of the amino acid Proline. Defects in PRODH are the cause of hyperprolinemia type 1, a disorder characterised by elevated serum proline levels. Defective PRODH may be involved in the psychiatric and behavioral phenotypes associated with the 22q11 velocardiofacial and DiGeorge syndrome and may be associated with susceptibility to schizophrenia 4 (SCZD4).

Type: Primary
Antigen: PRODH
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 141-240/600
Host:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR