Anti-CKS1B Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-5745R-A647
BOSSBS-5745R-A647EA 1190 CHF
BOSSBS-5745R-A647
Anti-CKS1B Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
CKS1 protein binds to the catalytic subunit of the cyclin dependent kinases and is essential for their biological function. The CKS1 mRNA is found to be expressed in different patterns through the cell cycle in HeLa cells, which reflects a specialized role for the encoded protein. At least two transcript variants have been identified for this gene, and it appears that only one of them encodes a protein.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CKS1B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR