Anti-RELA Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-5661R-CY5.5
BOSSBS-5661R-CY5.5EA 1240 CHF
BOSSBS-5661R-CY5.5
Anti-RELA Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
NF-kappa-B is a ubiquitous transcription factor involved in several biological processes. It is held in the cytoplasm in an inactive state by specific inhibitors. Upon degradation of the inhibitor, NF-kappa-B moves to the nucleus and activates transcription of specific genes. NF-kappa-B is composed of NFKB1 or NFKB2 bound to either REL, RELA, or RELB. The most abundant form of NF-kappa-B is NFKB1 complexed with the product of this gene, RELA. Four transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Sep 2011].
Recommended Dilutions: IF(IHC-P):More Product Information