Anti-SLC29A4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-5637R-HRP
BOSSBS-5637R-HRPEA 1800 CHF
BOSSBS-5637R-HRP
Anti-SLC29A4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
SLC29A4 (Solute carrier family 29 (nucleoside transporters), member 4) is involved in the re-uptake of monoamines into presynaptic neurons, thus determining the intensity and duration of monoamine neural signaling. It transports serotonin, dopamine and the neurotoxin 1-methyl-4-phenylpyridinium. It may also contribute to the regulation of extracellular adenosine concentrations.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: SLC29A4
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR