Anti-NFKBIA Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-5514R
BOSSBS-5514REA 956 CHF
BOSSBS-5514R
Anti-NFKBIA Rabbit Polyclonal Antibody
Antikörper
This gene encodes a member of the NF-kappa-B inhibitor family, which contain multiple ankrin repeat domains. The encoded protein interacts with REL dimers to inhibit NF-kappa-B/REL complexes which are involved in inflammatory responses. The encoded protein moves between the cytoplasm and the nucleus via a nuclear localization signal and CRM1-mediated nuclear export. Mutations in this gene have been found in ectodermal dysplasia anhidrotic with T-cell immunodeficiency autosomal dominant disease. [provided by RefSeq, Aug 2011].
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)