Anti-G3BP1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BS-5376R-A488
BOSSBS-5376R-A488EA 1200 CHF
BOSSBS-5376R-A488
Anti-G3BP1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
May be a regulated effector of stress granule assembly. Phosphorylation-dependent sequence-specific endoribonuclease in vitro. Cleaves exclusively between cytosine and adenine and cleaves MYC mRNA preferentially at the 3'-UTR. ATP- and magnesium-dependent helicase. Unwinds preferentially partial DNA and RNA duplexes having a 17 bp annealed portion and either a hanging 3' tail or hanging tails at both 5'- and 3'-ends. Unwinds DNA/DNA, RNA/DNA, and RNA/RNA substrates with comparable efficiency. Acts unidirectionally by moving in the 5' to 3' direction along the bound single-stranded DNA.