Anti-CARM1 Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-5285R-CY7
BOSSBS-5285R-CY7EA 1240 CHF
BOSSBS-5285R-CY7
Anti-CARM1 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Methylates (mono- and asymmetric dimethylation) the guanidino nitrogens of arginyl residues in several proteins involved in DNA packaging, transcription regulation, and mRNA stability. Recruited to promoters upon gene activation together with histone acetyltransferases from EP300/P300 and p160 families, methylates histone H3 at 'Arg-17' and activates transcription via chromatin remodeling. During nuclear hormone receptor activation and TCF7L2/TCF4 activation, acts synergically with EP300/P300 and either one of the p160 histone acetyltransferases NCOA1/SRC1, NCOA2/GRIP1 and NCOA3/ACTR orMore Product Information