Anti-PHKG1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-5009R-A488
BOSSBS-5009R-A488EA 1190 CHF
BOSSBS-5009R-A488
Anti-PHKG1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Phosphorylase b kinase catalyzes the phosphorylation of serine in certain substrates, including troponin I. Polymer of 16 chains, four each of alpha, beta, gamma, and delta. Alpha and beta are regulatory chains, gamma is the catalytic chain, and delta is calmodulin. The two calmodulin-binding domains appear to act in concert to bind a single molecule of calmodulin and are pseudosubstrate/autoinhibitory domains.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PHKG1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

TECHNISCHE DATEN

Antigen-Symbol PHKG1
Environmentally Preferable
Antigen-Name Phosphorylase Kinase, Gamma 1
Antigen-Synonyme PHKG1|Phkg|skeletal muscle isoform|PHKIN01|phosphorylase kinase gamma|phosphorylase b kinase gamma catalytic chain|phosphorylase kinase subunit gamma 1|gamma 1 muscle.|phosphorylase kinase
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Alexa Fluor® 488
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 5260
Isotyp IgG
ImmunoChemie Yes
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagertemperatur Store at 4°C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR