Anti-BDNF Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-4989R-A680
BOSSBS-4989R-A680EA 1200 CHF
BOSSBS-4989R-A680
Anti-BDNF Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
During development, promotes the survival and differentiation of selected neuronal populations of the peripheral and central nervous systems. Participates in axonal growth, pathfinding and in the modulation of dendritic growth and morphology. Major regulator of synaptic transmission and plasticity at adult synapses in many regions of the CNS. The versatility of BDNF is emphasized by its contribution to a range of adaptive neuronal responses including long-term potentiation (LTP), long-term depression (LTD), certain forms of short-term synaptic plasticity, as well as homeostatic regulation ofMore Product Information