Anti-ADORA1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-4235R-CY3
BOSSBS-4235R-CY3EA 1200 CHF
BOSSBS-4235R-CY3
Anti-ADORA1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Receptor for adenosine. The activity of this receptor is mediated by G proteins which inhibit adenylyl cyclase.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADORA1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

TECHNISCHE DATEN

Antigen-Symbol ADORA1
Antigen-Name Adenosine A1 Receptor
Environmentally Preferable
Antigen-Synonyme RDC7
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Cy3®
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 134
Isotyp IgG
ImmunoChemie Yes
Immunogen 147-176/326
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagertemperatur Store at 4°C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR