Anti-FANCG Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-4106R-A555
BOSSBS-4106R-A555EA 1190 CHF
BOSSBS-4106R-A555
Anti-FANCG Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
FANCG, involved in Fanconi anemia, confers resistance to both hygromycin and mitomycin C. FANCG contains a 5-prime GC-rich untranslated region characteristic of housekeeping genes. The putative 622-amino acid protein has a leucine-zipper motif at its N-terminus. Fanconi anemia is an autosomal recessive disorder with diverse clinical symptoms, including developmental anomalies, bone marrow failure, and early occurrence of malignancies. A minimum of 8 FA genes have been identified.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FANCG
Clonality: Polyclonal
Clone:
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR