Anti-SDHA Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-3970R-A680
BOSSBS-3970R-A680EA 1190 CHF
BOSSBS-3970R-A680
Anti-SDHA Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Flavoprotein (FP) subunit of succinate dehydrogenase (SDH) that is involved in complex II of the mitochondrial electron transport chain and is responsible for transferring electrons from succinate to ubiquinone (coenzyme Q). Can act as a tumor suppressor.

Type: Primary
Antigen: SDHA
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 561-664/664
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Order Now

TECHNISCHE DATEN

Antigen-Name SDHA
Environmentally Preferable
Antigen-Synonyme PGL5|CMD1GG|FP|Succinate dehydrogenase [ubiquinone] flavoprotein subunit, mitochondrial|SDHA|Flavoprotein subunit of complex II|SDHF|SDH1|SDH2
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Alexa Fluor® 680
Reaktivität Human,Mouse
Wirt Rabbit
Gen-ID 6389
Isotyp IgG
Western blot Yes
ImmunoChemie Yes
ImmunFluoreszenz Yes
Modification Unmodified
Epitop 561-664/664
Immunogen KLH conjugated synthetic peptide derived from human SDHA
Form Liquid
Molecular weight 70 kDa
Reinigung Purified by Protein A
Konzentration 1 µg/µl
Lagerungspuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagertemperatur Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Transporttemperatur 4 °C
Validierte Anwendungen ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR