Anti-PRKAR1B Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-3965R
BOSSBS-3965REA 965 CHF
BOSSBS-3965R
Anti-PRKAR1B Rabbit Polyclonal Antibody
Antikörper
cAMP is a signaling molecule important for a variety of cellular functions. cAMP exerts its effects by activating the cAMP-dependent protein kinase, which transduces the signal through phosphorylation of different target proteins. The inactive kinase holoenzyme is a tetramer composed of two regulatory and two catalytic subunits. cAMP causes the dissociation of the inactive holoenzyme into a dimer of regulatory subunits bound to four cAMP and two free monomeric catalytic subunits. Four different regulatory subunits and three catalytic subunits have been identified in humans. The proteinMore Product Information