Anti-NDUFA1 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-3956R
BOSSBS-3956REA 965 CHF
BOSSBS-3956R
Anti-NDUFA1 Rabbit Polyclonal Antibody
Antikörper
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: NDUFA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR