Anti-ADCY4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-3921R-A750
BOSSBS-3921R-A750EA 1190 CHF
BOSSBS-3921R-A750
Anti-ADCY4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
The ADCY4 gene encodes a member of the family of adenylate cyclases, which are membrane-associated enzymes that catalyse the formation of the secondary messenger cyclic adenosine monophosphate (cAMP). Mouse studies show that adenylate cyclase 4, along with adenylate cyclases 2 and 3, is expressed in olfactory cilia, suggesting that several different adenylate cyclases may couple to olfactory receptors and that there may be multiple receptor-mediated mechanisms for the generation of cAMP signals. Alternative splicing results in transcript variants.

Type: Primary
Antigen: ADCY4
Clonality:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR