Anti-CYP11B1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-3898R-CY5
BOSSBS-3898R-CY5EA 1180 CHF
BOSSBS-3898R-CY5
Anti-CYP11B1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Has steroid 11-beta-hydroxylase activity. In addition to this activity, the 18 or 19-hydroxylation of steroids and the aromatization of androstendione to estrone have also been ascribed to cytochrome P45 XIB.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CYP11B1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR