Anti-SIAH1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BS-3596R-A555
BOSSBS-3596R-A555EA 1190 CHF
BOSSBS-3596R-A555
Anti-SIAH1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
E3 ubiquitin-protein ligase that mediates ubiquitination and subsequent proteasomal degradation of target proteins. E3 ubiquitin ligases accept ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. Mediates E3 ubiquitin ligase activity either through direct binding to substrates or by functioning as the essential RING domain subunit of larger E3 complexes. Triggers the ubiquitin-mediated degradation of many substrates, including proteins involved in transcription regulation (ELL2, MYB, POU2AF1, PML andMore Product Information
Order Now
TECHNISCHE DATEN
Antigen-Symbol SIAH1
Antigen-Name Seven In Absentia Homolog 1
Antigen-Synonyme FLJ08065|SIAH1|seven in absentia homolog 1|hSIAH1|Siah-1a|seven in absentia homolog 1 (Drosophila)|sonic hedgehog homolog|Siah-1|HUMSIAH
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Alexa Fluor® 555
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 6477
Isotyp IgG
ImmunoChemie Yes
Immunogen 200-250/282
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagertemperatur Store at 4°C for 12 months
Transporttemperatur 4°C