Anti-T1 gamma Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-3582R-A750
BOSSBS-3582R-A750EA 1190 CHF
BOSSBS-3582R-A750
Anti-T1 gamma Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Acts as an E3 ubiquitin-protein ligase. Promotes SMAD4 ubiquitination, nuclear exclusion and degradation via the ubiquitin proteasome pathway. According to PubMed:16751102, does not promote a decrease in the level of endogenous SMAD4. May act as a transcriptional repressor. Inhibits the transcriptional response to TGF-beta/BMP signaling cascade. Plays a role in the control of cell proliferation. Its association with SMAD2 and SMAD3 stimulates erythroid differentiation of hematopoietic stem/progenitor (By similarity). Monoubiquitinates SMAD4 and acts as an inhibitor of SMAD4-dependent TGF-More Product Information
Order Now

TECHNISCHE DATEN

Environmentally Preferable
Antigen-Name T1 gamma
Antigen-Synonyme RET-fused gene 7 protein|Tripartite motif-containing protein 33|TIF1GAMMA|KIAA1113|TIFGAMMA|TF1G|E3 ubiquitin-protein ligase TRIM33|Protein Rfg7|TIF1G|Transcription intermediary factor 1-gamma|RFG7|TIF1-gamma|PTC7|Ectodermin homolog|ECTO|TRIM33
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Alexa Fluor® 750
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 51592
Isotyp IgG
ImmunoChemie Yes
ImmunFluoreszenz Yes
Durchflusszytometrie Yes
Modification Unmodified
Epitop 981-1127/1127
Immunogen KLH conjugated synthetic peptide derived from human TIF1 gamma
Form Liquid
Molecular weight 124 kDa
Reinigung Purified by Protein A
Konzentration 1 µg/µl
Lagerungspuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagertemperatur Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Transporttemperatur 4 °C
Validierte Anwendungen ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR