Anti-DLG2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Lieferant: Bioss
BS-3348R-A647
BOSSBS-3348R-A647EA 1230 CHF
BOSSBS-3348R-A647
Anti-DLG2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
PSD 93 is believed to participate in the clustering of certain proteins, including N-methyl-D-aspartate (NMDA) receptors and shaker-type potassium channels at the synaptic membrane. There are two principal modes of interaction between PSD 93 and other proteins. NMDA receptors and shaker-type potassium channels both share C-terminal sequence homology consisting of a threonine/serine-X-valine-COOH (T/SXV) motif. Other neuronal proteins that share this motif (beta 1 adrenergic receptor, some serotonin receptors, some sodium channel subunits, and additional potassium channel subunits) mayMore Product Information