Anti-DLG2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-3348R-A350
BOSSBS-3348R-A350EA 1230 CHF
BOSSBS-3348R-A350
Anti-DLG2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
PSD 93 is believed to participate in the clustering of certain proteins, including N-methyl-D-aspartate (NMDA) receptors and shaker-type potassium channels at the synaptic membrane. There are two principal modes of interaction between PSD 93 and other proteins. NMDA receptors and shaker-type potassium channels both share C-terminal sequence homology consisting of a threonine/serine-X-valine-COOH (T/SXV) motif. Other neuronal proteins that share this motif (beta 1 adrenergic receptor, some serotonin receptors, some sodium channel subunits, and additional potassium channel subunits) mayMore Product Information
Order Now

TECHNISCHE DATEN

Antigen-Symbol DLG2
Environmentally Preferable
Antigen-Name Discs, Large Homolog 2 (Drosophila)
Antigen-Synonyme Postsynaptic density protein PSD-93Channel-associated protein of synapse-110|PPP1R58|PSD93,Chapsyn-110|Chapsyn-1|PSD-93|Post Synaptic Density 93|Discs large homolog 2
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Alexa Fluor® 350
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 1740
Isotyp IgG
ImmunoChemie Yes
Modification phospho-Tyr340
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagertemperatur Store at 4°C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR