Anti-NPM Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-3308R
BOSSBS-3308REA 989 CHF
BOSSBS-3308R
Anti-NPM Rabbit Polyclonal Antibody
Antikörper
This gene encodes a phosphoprotein which moves between the nucleus and the cytoplasm. The gene product is thought to be involved in several processes including regulation of the ARF/p53 pathway. A number of genes are fusion partners have been characterized, in particular the anaplastic lymphoma kinase gene on chromosome 2. Mutations in this gene are associated with acute myeloid leukemia. More than a dozen pseudogenes of this gene have been identified. Alternative splicing results in multiple transcript variants.[provided by RefSeq, Nov 2009].

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:More Product Information
Order Now

TECHNISCHE DATEN

Antigen-Symbol NPM
Antigen-Name Nucleophosmin
Environmentally Preferable
Antigen-Synonyme MGC104254|NO38|Nucleophosmin|Nucleolar protein NO38|numatrin|B23|Nucleolar phosphoprotein B23|NPM1
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 4869
Isotyp IgG
Western blot Yes
ImmunoChemie Yes
Modification Ser4
Form liquid
Reinigung Purified by Protein A.
Konzentration 1 ug/ul
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Transporttemperatur 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR