Anti-PTK2 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-3162R
BOSSBS-3162REA 989 CHF
BOSSBS-3162R
Anti-PTK2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PTK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR