Anti-BCL2 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-3052R-CY5.5
BOSSBS-3052R-CY5.5EA 1240 CHF
BOSSBS-3052R-CY5.5
Anti-BCL2 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Suppresses apoptosis in a variety of cell systems including factor-dependent lymphohematopoietic and neural cells. Regulates cell death by controlling the mitochondrial membrane permeability. Appears to function in a feedback loop system with caspases. Inhibits caspase activity either by preventing the release of cytochrome c from the mitochondria and/or by binding to the apoptosis-activating factor (APAF-1).

Recommended Dilutions: IF(IHC-P): 1:50-200; IF(ICC)(1:50-200

Type: Primary
Antigen: BCL2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR