Anti-MAP3K5 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-3031R
BOSSBS-3031REA 992 CHF
BOSSBS-3031R
Anti-MAP3K5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAP3K5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR